Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00476769_s1
          See other PHXR2 GE Assays ›
          Gene Symbol:
          Phxr2
          Gene Name:
          per-hexamer repeat gene 2
          Gene Aliases:
          SP28
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00476769_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phxr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC119390.1 - 48 8734139 71
          BC119392.1 - 48 8734141 71
          X12808.1 - 52 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002759.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC119390.1,BC119392.1,X12808.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076458 BC119392.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076458 X12808.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076458 BC119390.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076458 BC119392.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076458 X12808.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076458 BC119390.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27385160 - -
          Whole Mouse Genome G4122A A_51_P207883 X12808.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109763 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10372112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10372113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10372111 - -
          Affymetrix HT MG-430 PM Array Plate 1422287_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10372110 - -
          Mouse Genome 430 2.0 Array 1422287_at - -
          Mouse Genome 430 2.0 Array 1422287_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000021080.mm.1 TC1000002759.mm.1 MTA TC
          Mouse Expression Set 430 1422287_PM_at - -
          Mouse Expression Set 430 1422287_at - -
          Murine Genome U74v2 Set 101699_at - -
          Murine Genome U74v2 Set 101699_PM_at - -
          Mouse Genome 430A 2.0 Array 1422287_at - -
          Mouse Genome 430A 2.0 Array 1422287_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422287_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422287_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 674606 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3520273 - -
          Illumina mouse-6 v1.0 expression beadchip 3520273 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003520273 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2604002 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2604002 - -
          mouse-6 v1.1 (Illumina) scl37566.1.1_140-S - -
          Illumina Sentrix mouse-6.v1 bead array 3520273 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37566.1.1_140-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3520273 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2604002 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline