Search
Search
| Gene Symbol: Rrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010925.2 | NP_035055.2 | 8 - 9 | 1070 | 75 | |
| GenBank mRNA | AF312394.1 | - | 7 - 8 | 833 | 75 | |
| AK077540.1 | - | 8 - 9 | 995 | 75 | ||
| BC003913.1 | - | 1 - 2 | 166 | 75 | ||
| BC022910.1 | - | 7 - 8 | 936 | 4165783 | 75 | |
| BC065993.1 | - | 8 - 9 | 1032 | 5684672 | 75 | |
| BC065994.1 | - | 8 - 9 | 990 | 5685269 | 75 | |
| U79773.1 | - | 9 - 10 | 869 | 75 | ||
| U79774.1 | - | 8 - 9 | 1022 | 75 | ||
Set Membership: |
|