Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00478333_g1
          See other PSAP GE Assays ›
          Gene Symbol:
          Psap
          Gene Name:
          prosaposin
          Gene Aliases:
          AI037048, SGP-1
          Chromosome Location:
          Chr.10: 60277628 - 60302600 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00478333_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001146120.1 NP_001139592.1 8 - 9 1024 78
          NM_001146121.1 NP_001139593.1 9 - 10 1030 78
          NM_001146123.1 NP_001139595.1 8 - 9 997 78
          NM_001146124.1 NP_001139596.1 8 - 9 940 78
          NM_011179.3 NP_035309.3 9 - 10 1033 78
          GenBank mRNA AK088369.1 - 8 - 9 938 78
          AK089998.1 - 8 - 9 942 78
          AK093881.1 - 8 - 9 938 78
          AK135637.1 - 8 - 9 938 78
          AK145607.1 - 8 - 9 945 78
          AK145802.1 - 8 - 9 939 78
          AK148152.1 - 8 - 9 965 78
          AK148545.1 - 9 - 10 949 78
          AK149785.1 - 8 - 9 941 78
          AK150331.1 - 8 - 9 940 78
          AK150645.1 - 8 - 9 938 78
          AK151034.1 - 8 - 9 938 78
          AK151173.1 - 8 - 9 939 78
          AK151230.1 - 8 - 9 938 78
          AK151234.1 - 8 - 9 913 78
          AK151251.1 - 8 - 9 911 78
          AK151525.1 - 8 - 9 940 78
          AK151529.1 - 8 - 9 913 78
          AK151686.1 - 8 - 9 974 78
          AK151852.1 - 8 - 9 940 78
          AK151875.1 - 8 - 9 937 78
          AK151891.1 - 8 - 9 938 78
          AK151907.1 - 8 - 9 940 78
          AK151943.1 - 8 - 9 940 78
          AK152117.1 - 8 - 9 940 78
          AK152126.1 - 8 - 9 938 78
          AK152134.1 - 8 - 9 940 78
          AK152203.1 - 8 - 9 972 78
          AK152280.1 - 8 - 9 938 78
          AK152319.1 - 8 - 9 942 78
          AK152355.1 - 8 - 9 940 78
          AK152408.1 - 8 - 9 943 78
          AK152505.1 - 8 - 9 940 78
          AK152557.1 - 8 - 9 940 78
          AK152605.1 - 8 - 9 940 78
          AK152819.1 - 8 - 9 952 78
          AK153404.1 - 8 - 9 938 78
          AK159108.1 - 8 - 9 968 78
          AK159167.1 - 8 - 9 939 78
          AK159198.1 - 8 - 9 938 78
          AK159266.1 - 8 - 9 938 78
          AK159333.1 - 8 - 9 938 78
          AK159341.1 - 8 - 9 938 78
          AK159384.1 - 8 - 9 938 78
          AK159414.1 - 8 - 9 941 78
          AK159427.1 - 8 - 9 940 78
          AK159451.1 - 8 - 9 938 78
          AK159457.1 - 8 - 9 952 78
          AK159463.1 - 8 - 9 940 78
          AK159487.1 - 8 - 9 938 78
          AK159510.1 - 8 - 9 938 78
          AK159528.1 - 8 - 9 938 78
          AK159530.1 - 8 - 9 938 78
          AK159589.1 - 8 - 9 940 78
          AK159604.1 - 8 - 9 938 78
          AK159622.1 - 7 - 8 830 78
          AK159634.1 - 8 - 9 938 78
          AK159638.1 - 8 - 9 939 78
          AK159657.1 - 8 - 9 938 78
          AK159664.1 - 8 - 9 938 78
          AK159685.1 - 8 - 9 938 78
          AK159692.1 - 8 - 9 938 78
          AK159709.1 - 8 - 9 938 78
          AK159720.1 - 8 - 9 938 78
          AK159722.1 - 8 - 9 938 78
          AK159728.1 - 8 - 9 940 78
          AK159840.1 - 8 - 9 938 78
          AK159861.1 - 8 - 9 938 78
          AK159862.1 - 8 - 9 940 78
          AK159877.1 - 8 - 9 938 78
          AK159910.1 - 8 - 9 938 78
          AK159915.1 - 8 - 9 938 78
          AK159936.1 - 8 - 9 940 78
          AK159982.1 - 8 - 9 938 78
          AK159994.1 - 8 - 9 938 78
          AK160015.1 - 8 - 9 938 78
          AK160019.1 - 8 - 9 940 78
          AK160021.1 - 8 - 9 938 78
          AK160024.1 - 8 - 9 938 78
          AK160046.1 - 8 - 9 938 78
          AK160049.1 - 8 - 9 940 78
          AK160073.1 - 8 - 9 938 78
          AK160108.1 - 8 - 9 938 78
          AK160877.1 - 8 - 9 941 78
          AK161313.1 - 8 - 9 955 78
          AK166700.1 - 8 - 9 938 78
          AK166707.1 - 8 - 9 937 78
          AK166744.1 - 8 - 9 936 78
          AK166811.1 - 8 - 9 938 78
          AK166849.1 - 8 - 9 938 78
          AK166879.1 - 8 - 9 940 78
          AK166909.1 - 8 - 9 938 78
          AK166920.1 - 8 - 9 938 78
          AK166946.1 - 8 - 9 943 78
          AK166959.1 - 8 - 9 938 78
          AK166984.1 - 8 - 9 938 78
          AK167014.1 - 8 - 9 938 78
          AK167025.1 - 8 - 9 938 78
          AK167073.1 - 8 - 9 938 78
          AK167099.1 - 8 - 9 941 78
          AK167108.1 - 8 - 9 938 78
          AK167111.1 - 8 - 9 938 78
          AK167147.1 - 8 - 9 938 78
          AK167198.1 - 8 - 9 945 78
          AK167213.1 - 8 - 9 938 78
          AK167237.1 - 8 - 9 938 78
          AK167243.1 - 8 - 9 939 78
          AK167252.1 - 8 - 9 940 78
          AK167418.1 - 8 - 9 940 78
          AK167719.1 - 8 - 9 940 78
          AK167902.1 - 8 - 9 1008 78
          AK170416.1 - 8 - 9 937 78
          AK182197.1 - 2 - 3 175 78
          AK195069.1 - 2 - 3 181 78
          BC036266.1 - 7 - 8 804 78
          BC141089.1 - 9 - 10 943 9055701 78
          BC145359.1 - 8 - 9 937 9053917 78
          S36200.1 - 9 - 10 923 78
          U27340.1 - 8 - 9 938 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000547.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psap

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          sphingolipid metabolic process
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          regulation of lipid metabolic process
          regulation of MAPK cascade
          positive regulation of MAPK cascade
          developmental growth
          prostate gland growth
          epithelial cell differentiation involved in prostate gland development
          cellular response to organic substance
          negative regulation of hydrogen peroxide-induced cell death
          G-protein coupled receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000003706.mm.1 NM_001146120.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003706.mm.1 NM_001146120.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003706.mm.1 NM_001146121.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003706.mm.1 NM_001146121.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003706.mm.1 NM_001146122.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003706.mm.1 NM_001146122.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003706.mm.1 NM_001146123.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003706.mm.1 NM_001146123.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003706.mm.1 NM_001146124.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003706.mm.1 NM_001146124.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003706.mm.1 NM_011179.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003706.mm.1 NM_011179.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003706.mm.1 TC1000000547.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline