Search
Search
| Gene Symbol: Nek4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF099067.1 | - | 3 - 4 | 701 | 83 | |
| AJ223071.1 | - | 3 - 4 | 791 | 83 | ||
| AK078809.1 | - | 3 - 4 | 813 | 83 | ||
| AK132803.1 | - | 3 - 4 | 729 | 83 | ||
| AK144130.1 | - | 3 - 4 | 675 | 83 | ||
| AK147381.1 | - | 3 - 4 | 779 | 83 | ||
| AK149584.1 | - | 3 - 4 | 729 | 83 | ||
| AK168094.1 | - | 3 - 4 | 712 | 83 | ||
| BC057939.1 | - | 3 - 4 | 703 | 5325558 | 83 | |
| Y09234.1 | - | 3 - 4 | 791 | 83 | ||
Set Membership: |
|