| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28941738 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28941739 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28941740 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28941742 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2000683 |
AK009478.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2000683 |
AK170778.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2000683 |
BQ176032.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2000683 |
NM_001110331.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2000683 |
NM_001110332.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
390673 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
33283 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
342960 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
150422 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
830042 |
AK035561.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
830042 |
NM_172524.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
297658 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
916138 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
477020 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
695465 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
7115 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
37974 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P144747 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P107588 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2167776 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2000683 |
AK009478.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2000683 |
AK170778.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2000683 |
BQ176032.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2000683 |
NM_001110331.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2000683 |
NM_001110332.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P394665 |
AK009478.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P394665 |
AK170778.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P394665 |
BQ176032.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P394665 |
NM_001110331.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P394665 |
NM_001110332.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941748 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941726 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941735 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941745 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941773 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941757 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941731 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941749 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941751 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941738 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941737 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941768 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941772 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941727 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941740 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941729 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941741 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941742 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941774 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941733 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941753 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941758 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941744 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941766 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941754 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941756 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941730 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941769 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941763 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28941760 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07834955 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07834987 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P144747 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P107588 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P394665 |
AK009478.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P394665 |
AK170778.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P394665 |
BQ176032.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P394665 |
NM_001110331.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P394665 |
NM_001110332.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P107588 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P144747 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10754218 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10754215 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10754209 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10754214 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10754210 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28941739 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28941740 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28941738 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28941742 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941702 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941712 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941636 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941595 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941730 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941654 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941780 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941705 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941600 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941615 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941742 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941586 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941648 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941740 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941605 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941699 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941744 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941785 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941639 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941741 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941599 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941763 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941766 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941769 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941641 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941776 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941682 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941675 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941622 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941650 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941592 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941584 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941745 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941676 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941656 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941729 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941603 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941660 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941629 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941778 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941633 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941754 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941630 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941685 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941617 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941657 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941733 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941697 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941782 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941709 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941704 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941694 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941710 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941591 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941631 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941720 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941735 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941651 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941723 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941738 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941751 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941608 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941786 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941628 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941652 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941607 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941757 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941768 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941679 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941737 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941637 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941772 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941618 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941655 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28941731 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941691 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941745 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941697 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941708 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941700 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941749 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941730 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10804599 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941731 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941758 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941740 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941712 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941709 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941748 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10804603 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941720 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941729 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941713 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941664 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941710 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941742 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941754 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11040895 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11102148 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941757 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941773 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941733 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941667 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941753 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941727 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941750 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941699 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941694 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941725 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941762 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10974047 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941774 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941760 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941705 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941763 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941769 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941741 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941723 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10667605 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941738 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10754219 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941744 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941768 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28941702 |
- |
- |