Search
Search
| Gene Symbol: Miox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019977.2 | NP_064361.2 | 1 - 2 | 46 | 80 | |
| GenBank mRNA | AF197127.1 | - | 1 - 2 | 44 | 80 | |
| AK002406.1 | - | 1 - 2 | 60 | 80 | ||
| BC013543.1 | - | 1 - 2 | 46 | 4242159 | 80 | |
Set Membership: |
|