Search
Search

Pcbd1| Gene Symbol: Pcbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025273.4 | NP_079549.1 | 1 - 2 | 88 | 57 | |
| GenBank mRNA | AK007401.1 | - | 1 - 2 | 60 | 57 | |
| BY294059.1 | - | 1 - 2 | 76 | 57 | ||
Set Membership: |
|