Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481245_m1
          See other JOSD2 GE Assays ›
          Gene Symbol:
          Josd2
          Gene Name:
          Josephin domain containing 2
          Gene Aliases:
          1110007C05Rik
          Chromosome Location:
          Chr.7: 44467980 - 44471658 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481245_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Josd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001205070.1 NP_001191999.1 3 - 4 484 92
          NM_001205071.1 NP_001192000.1 2 - 3 372 92
          NM_001205072.1 NP_001192001.1 2 - 3 341 92
          NM_001205073.1 NP_001192002.1 2 - 3 338 92
          NM_025368.4 NP_079644.1 2 - 3 369 92
          GenBank mRNA AK002872.1 - 2 - 3 210 92
          AK003525.1 - 2 - 3 294 92
          BC018244.1 - 3 - 4 313 4224952 92
          CA326008.1 - 2 - 3 283 92

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Josd2

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          protein deubiquitination
          thiol-dependent ubiquitin-specific protease activity
          peptidase activity
          omega peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25087597 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25087598 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25087599 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2055875 NM_001205073.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2055875 NM_025368.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2055875 AK002872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2055875 AK003525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2055875 NM_001205070.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 811526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96262 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121999 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131225 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158060 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2055875 NM_001205073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2055875 NM_025368.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2055875 AK002872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2055875 AK003525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2055875 NM_001205070.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25087597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25087606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25087615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25087600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25087611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25087603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25087613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25087601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25087612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25087608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25087602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25087610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25087604 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158060 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131225 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121999 - -
          Whole Mouse Genome G4122A A_51_P325491 NM_001205073.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325491 NM_025368.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325491 AK002872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325491 AK003525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P325491 AK189520.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130320 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158060 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131225 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121999 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06041201 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06041206 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25087599 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25087598 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25087597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25087573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06041195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06295312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06256241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06295320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06041210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06295314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06041199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06256242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06256239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25087617 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552684 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552686 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552683 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552685 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552687 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552682 - -
          Affymetrix HT MG-430 PM Array Plate 1449046_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552681 - -
          Mouse Genome 430 2.0 Array 1449046_a_at - -
          Mouse Genome 430 2.0 Array 1449046_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66124_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66124_PM_at - -
          Mouse Expression Set 430 1449046_a_at - -
          Mouse Expression Set 430 1449046_PM_a_at - -
          Murine Genome U74v2 Set 95041_at - -
          Murine Genome U74v2 Set 95041_PM_at - -
          Mouse Genome 430A 2.0 Array 1449046_a_at - -
          Mouse Genome 430A 2.0 Array 1449046_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449046_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449046_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5080440 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005080440 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2667614 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2667614 - -
          mouse-6 v1.1 (Illumina) scl32713.7.1_59-S - -
          Illumina Sentrix mouse-6.v1 bead array 5080440 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32713.7.1_59-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5080440 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2667614 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2667614 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl32713.7.1_59-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline