Search
Search
| Gene Symbol: Aig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025446.3 | NP_079722.1 | 1 - 2 | 214 | 64 | |
| GenBank mRNA | AK005332.1 | - | 1 - 2 | 242 | 64 | |
| AK008210.1 | - | 1 - 2 | 217 | 64 | ||
| AK084050.1 | - | 1 - 2 | 360 | 64 | ||
| AK162306.1 | - | 1 - 2 | 233 | 64 | ||
| BC035305.1 | - | 1 - 2 | 364 | 3980923 | 64 | |
Set Membership: |
|