Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481391_m1
          See other EAPP GE Assays ›
          Gene Symbol:
          Eapp
          Gene Name:
          E2F-associated phosphoprotein
          Gene Aliases:
          1810011O16Rik
          Chromosome Location:
          Chr.12: 54670340 - 54695896 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481391_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eapp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025456.3 NP_079732.1 5 - 6 661 57
          GenBank mRNA AK007442.1 - 5 - 6 616 57
          AK145027.1 - 5 - 6 680 57
          AK145041.1 - 5 - 6 680 57
          AY882557.1 - 5 - 6 580 57
          BC037622.1 - 5 - 6 688 5373275 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001749.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eapp

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          positive regulation of transcription elongation from RNA polymerase II promoter
          regulation of transcription elongation from RNA polymerase II promoter
          negative regulation of transcription elongation from RNA polymerase II promoter
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28449068 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28449065 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28449066 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28449069 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28449067 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P282424 BC037622.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P282424 AK145027.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P282424 AK145041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P282424 AY882557.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P282424 NM_025456.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P282424 BC037622.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P282424 AK145027.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P282424 AK145041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P282424 AY882557.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P282424 NM_025456.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017312 BC037622.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017312 AK145027.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017312 AK145041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017312 AY882557.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017312 NM_025456.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111009 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 88561 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292986 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377429 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 804580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55264 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 281323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950622 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 87330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 74584 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51523 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148861 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180206 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P282424 BC037622.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P282424 AK145027.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P282424 AK145041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P282424 AY882557.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P282424 NM_025456.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P282424 BC037622.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P282424 AK145027.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P282424 AK145041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P282424 AY882557.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P282424 NM_025456.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017312 BC037622.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017312 AK145027.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017312 AK145041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017312 AY882557.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017312 NM_025456.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28449062 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07626060 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07626080 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148861 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180206 - -
          Whole Mouse Genome G4122A A_51_P282424 BC037622.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P282424 AK145027.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P282424 AK145041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P282424 AY882557.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P282424 NM_025456.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P282424 BC037622.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P282424 AK145027.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P282424 AK145041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P282424 AY882557.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P282424 NM_025456.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116817 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148861 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180206 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09966304 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09966301 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09966306 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09966298 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09966294 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09966307 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28449066 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28449065 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28449068 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28449067 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28449069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28448956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28449064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09779088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28448974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28448980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28448981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28448971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28448969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10071583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28448970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10072392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28448977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10031843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28448972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28448975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10179758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09966294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10173062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09779087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10184813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10010744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28448979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09967884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09750461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28449043 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10400328 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400330 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400331 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400332 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400333 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400329 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400327 - -
          Affymetrix HT MG-430 PM Array Plate 1432444_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10400326 - -
          Mouse Genome 430 2.0 Array 1432444_PM_a_at - -
          Mouse Genome 430 2.0 Array 1432444_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66266_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66266_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200012013.mm.1 NM_025456.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200012013.mm.1 TC1200001749.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200012012.mm.1 NM_025456.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200012012.mm.1 TC1200001749.mm.1 MTA TC
          Mouse Expression Set 430 1432444_a_at - -
          Mouse Expression Set 430 1432444_PM_a_at - -
          Murine Genome U74v2 Set 165695_PM_at - -
          Murine Genome U74v2 Set 165695_at - -
          Mouse Genome 430A 2.0 Array 1432444_a_at - -
          Mouse Genome 430A 2.0 Array 1432444_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432444_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432444_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 621682 - -
          ABI Mouse Genome Survey Microarray 872563 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4070575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004070575 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258998 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258998 - -
          mouse-6 v1.1 (Illumina) scl42463.7.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 4070575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42463.7.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4070575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1258998 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258998 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl42463.7.1_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline