Search
Search
| Gene Symbol: Cap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301067.1 | NP_001287996.1 | 8 - 9 | 1063 | 92 | |
| NM_007598.4 | NP_031624.2 | 8 - 9 | 1047 | 92 | ||
| GenBank mRNA | AK053351.1 | - | 8 - 9 | 871 | 92 | |
| AK128889.1 | - | 8 - 9 | 869 | 92 | ||
| AK137222.1 | - | 8 - 9 | 852 | 92 | ||
| AK144094.1 | - | 8 - 9 | 855 | 92 | ||
| AK154002.1 | - | 8 - 9 | 1067 | 92 | ||
| AK156556.1 | - | 8 - 9 | 881 | 92 | ||
| AK158150.1 | - | 7 - 8 | 1711 | 92 | ||
| AK165565.1 | - | 8 - 9 | 850 | 92 | ||
| AK169652.1 | - | 8 - 9 | 842 | 92 | ||
| AK170901.1 | - | 8 - 9 | 858 | 92 | ||
| AK171388.1 | - | 8 - 9 | 865 | 92 | ||
| BC005446.1 | - | 7 - 8 | 846 | 3600514 | 92 | |
| BC005472.1 | - | 7 - 8 | 839 | 3590890 | 92 | |
| L12367.1 | - | 8 - 9 | 873 | 92 | ||
Set Membership: |
|