Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00483050_m1
          See other CTNND1 GE Assays ›
          Gene Symbol:
          Ctnnd1
          Gene Name:
          catenin (cadherin associated protein), delta 1
          Gene Aliases:
          AA409437, AU019353, Catns, Ctnnd, P120, mKIAA0384
          Chromosome Location:
          Chr.2: 84600781 - 84658972 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00483050_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctnnd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085448.1 NP_001078917.1 11 - 12 2391 89
          NM_001085449.1 NP_001078918.1 9 - 10 2084 89
          NM_001085450.1 NP_001078919.1 11 - 12 2391 89
          NM_001085453.1 NP_001078922.1 10 - 11 2143 89
          NM_007615.4 NP_031641.2 12 - 13 2509 89
          GenBank mRNA AB093237.1 - 12 - 13 2225 89
          AK133193.1 - 11 - 12 2112 89
          AK161790.1 - 12 - 13 2285 89
          AK164485.1 - 11 - 12 2376 89
          AK168453.1 - 9 - 10 2082 89
          BC043108.1 - 11 - 12 2326 6406902 89
          BC046589.1 - 9 - 10 1884 89
          BC054544.1 - 12 - 13 2442 6408956 89
          Z17804.1 - 2286 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003869.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctnnd1

          Panther Classification:

          Molecular Function -

          intermediate filament binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of a polarized epithelium
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cell adhesion
          salivary gland morphogenesis
          Wnt signaling pathway
          single organismal cell-cell adhesion
          keratinocyte differentiation
          epithelial cell differentiation involved in salivary gland development
          negative regulation of canonical Wnt signaling pathway
          receptor binding
          protein binding
          protein kinase binding
          protein phosphatase binding
          protein domain specific binding
          cadherin binding
          cell adhesion molecule binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200033455.mm.1 NM_001085448.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033455.mm.1 NM_001085448.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033455.mm.1 NM_001085449.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033455.mm.1 NM_001085449.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033455.mm.1 NM_001085450.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033455.mm.1 NM_001085450.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033455.mm.1 NM_001085453.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033455.mm.1 NM_007615.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033455.mm.1 NM_007615.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033455.mm.1 TC0200003869.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline