| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600000853.mm.1 |
NM_001243064.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000853.mm.1 |
NM_001243064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000853.mm.1 |
NM_007616.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000853.mm.1 |
NM_007616.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000853.mm.1 |
TC0600000128.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000855.mm.1 |
NM_001243064.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000855.mm.1 |
NM_001243064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000855.mm.1 |
NM_007616.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000855.mm.1 |
NM_007616.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000855.mm.1 |
TC0600000128.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000857.mm.1 |
NM_001243064.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000857.mm.1 |
NM_001243064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000857.mm.1 |
NM_007616.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000857.mm.1 |
NM_007616.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000857.mm.1 |
TC0600000128.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000845.mm.1 |
NM_001243064.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000845.mm.1 |
NM_001243064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000845.mm.1 |
TC0600000128.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000856.mm.1 |
NM_001243064.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000856.mm.1 |
NM_001243064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000856.mm.1 |
NM_007616.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000856.mm.1 |
NM_007616.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000856.mm.1 |
TC0600000128.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000846.mm.1 |
NM_001243064.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000846.mm.1 |
NM_001243064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000846.mm.1 |
TC0600000128.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000854.mm.1 |
NM_001243064.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000854.mm.1 |
NM_001243064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000854.mm.1 |
NM_007616.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000854.mm.1 |
NM_007616.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000854.mm.1 |
TC0600000128.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000858.mm.1 |
NM_001243064.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000858.mm.1 |
NM_001243064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000858.mm.1 |
NM_007616.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000858.mm.1 |
NM_007616.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000858.mm.1 |
TC0600000128.mm.1 |
MTA TC |