Search
Search
| Gene Symbol: Cdh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007666.3 | NP_031692.2 | 10 - 11 | 1894 | 111 | |
| GenBank mRNA | BC107008.1 | - | 9 - 10 | 1635 | 40047565 | 111 |
| BC107013.1 | - | 9 - 10 | 1635 | 40048308 | 111 | |
| D82029.1 | - | 9 - 10 | 1635 | 111 | ||
| U67399.1 | - | 2 - 3 | 202 | 111 | ||
Set Membership: |
|