Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00483284_m1
          See other CHAD GE Assays ›
          Gene Symbol:
          Chad
          Gene Name:
          chondroadherin
          Gene Aliases:
          SLRR4A
          Chromosome Location:
          Chr.11: 94565074 - 94569127 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00483284_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chad
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007689.4 NP_031715.1 1 - 2 795 59
          GenBank mRNA AK158297.1 - 1 - 2 823 59
          AK160489.1 - 1 - 2 391 59
          BC012672.1 - 1 - 2 834 4192855 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001439.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chad

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s)

          bone development
          negative regulation of bone trabecula formation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P480073 NM_007689.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P480073 AK158297.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P480073 AK160489.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P480073 BC012672.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P480073 NM_007689.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P480073 AK158297.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P480073 AK160489.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P480073 BC012672.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159302 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P480073 NM_007689.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P480073 AK158297.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P480073 AK160489.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P480073 BC012672.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P480073 NM_007689.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P480073 AK158297.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P480073 AK160489.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P480073 BC012672.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28033707 - -
          Whole Mouse Genome G4122A A_51_P480073 NM_007689.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P480073 AK158297.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P480073 AK160489.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P480073 BC012672.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P480073 NM_007689.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P480073 AK158297.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P480073 AK160489.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P480073 BC012672.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100823 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102460 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09449794 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09449789 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09449787 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09449773 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09449760 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09449782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28033718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09449786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09449788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09449798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09185328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09368821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09417100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09449763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09082847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09368822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09256493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08999942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09417097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28033689 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10380402 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380400 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380399 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380401 - -
          Affymetrix HT MG-430 PM Array Plate 1420569_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10380398 - -
          Mouse Genome 430 2.0 Array 1420569_PM_at - -
          Mouse Genome 430 2.0 Array 1420569_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12643_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12643_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100013358.mm.1 NM_007689.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013358.mm.1 NM_007689.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013358.mm.1 TC1100001439.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100013359.mm.1 NM_007689.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013359.mm.1 NM_007689.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013359.mm.1 TC1100001439.mm.1 MTA TC
          Mouse Expression Set 430 1420569_PM_at - -
          Mouse Expression Set 430 1420569_at - -
          Murine Genome U74v2 Set 99913_at - -
          Murine Genome U74v2 Set 99913_PM_at - -
          Mouse Genome 430A 2.0 Array 1420569_at - -
          Mouse Genome 430A 2.0 Array 1420569_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420569_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420569_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 655456 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3060100 - -
          Illumina mouse-6 v1.0 expression beadchip 3060100 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060100 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2659340 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2659340 - -
          mouse-6 v1.1 (Illumina) scl41026.4.1_104-S - -
          Illumina Sentrix mouse-6.v1 bead array 3060100 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41026.4.1_104-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060100 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2659340 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2659340 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline