Search
Search
| Gene Symbol: Cmah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111110.2 | NP_001104580.1 | 10 - 11 | 1448 | 109 | |
| NM_001284519.1 | NP_001271448.1 | 11 - 12 | 1610 | 109 | ||
| NM_007717.5 | NP_031743.3 | 9 - 10 | 1103 | 109 | ||
| GenBank mRNA | AB061276.1 | - | 10 - 11 | 1432 | 109 | |
| AB061277.1 | - | 11 - 12 | 1594 | 109 | ||
| AK145110.1 | - | 8 - 9 | 110 | |||
| AK145199.1 | - | 9 - 10 | 1104 | 109 | ||
| AK166486.1 | - | 9 - 10 | 1104 | 109 | ||
| BC028474.1 | - | 10 - 11 | 1458 | 109 | ||
| D21826.1 | - | 8 - 9 | 1075 | 109 | ||
Set Membership: |
|