Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00484387_s1
          See other FOXD4 GE Assays ›
          Gene Symbol:
          Foxd4
          Gene Name:
          forkhead box D4
          Gene Aliases:
          FREAC5, Fkh2
          Chromosome Location:
          Chr.19: 24898965 - 24901309 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00484387_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Foxd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008022.2 NP_032048.1 1 2233 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001288.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Foxd4

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          anatomical structure morphogenesis
          cell differentiation
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026434 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026431 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026409 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026412 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026439 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026436 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026433 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026410 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026429 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026430 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026413 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026435 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2160820 NM_008022.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2160820 BC138432.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2160820 BC138434.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 110105 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101715 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2160820 NM_008022.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2160820 BC138432.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2160820 BC138434.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P443584 NM_008022.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32026461 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101715 - -
          Whole Mouse Genome G4122A A_51_P443584 NM_008022.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126210 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101715 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14271252 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14271249 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14271246 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14271245 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14271258 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14271248 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14271259 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14271260 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14271247 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026435 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026413 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026412 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026434 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026429 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026436 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026410 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026439 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026409 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026430 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026433 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32026431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32026394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14364580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14143336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14361557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14225823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14361540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14361559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14397531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14361539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14271258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14217757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32026393 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466816 - -
          Affymetrix HT MG-430 PM Array Plate 1422318_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10466814 - -
          Mouse Genome 430 2.0 Array 1422318_PM_at - -
          Mouse Genome 430 2.0 Array 1422318_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14237_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14237_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900011610.mm.1 NM_008022.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011610.mm.1 NM_008022.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011610.mm.1 TC1900001288.mm.1 MTA TC
          Mouse Expression Set 430 1422318_PM_at - -
          Mouse Expression Set 430 1422318_at - -
          Murine Genome U74v2 Set 101648_at - -
          Murine Genome U74v2 Set 101648_PM_at - -
          Mouse Genome 430A 2.0 Array 1422318_PM_at - -
          Mouse Genome 430A 2.0 Array 1422318_at - -
          Affymetrix HT Mouse Genome 430A Array 1422318_at - -
          Affymetrix HT Mouse Genome 430A Array 1422318_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 398166 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2470315 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002470315 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2744317 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2744317 - -
          mouse-6 v1.1 (Illumina) scl6077.1.1_249-S - -
          Illumina Sentrix mouse-6.v1 bead array 2470315 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl6077.1.1_249-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2470315 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2744317 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2744317 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline