Search
Search
| Gene Symbol: Fmr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290424.1 | NP_001277353.1 | 4 - 5 | 541 | 110 | |
| NM_008031.3 | NP_032057.2 | 4 - 5 | 541 | 110 | ||
| GenBank mRNA | AF461114.1 | - | 4 - 5 | 265 | 110 | |
| AK046605.1 | - | 4 - 5 | 501 | 110 | ||
| AK053211.1 | - | 1 - 2 | 283 | 110 | ||
| AK053701.1 | - | 4 - 5 | 501 | 110 | ||
| AK053829.1 | - | 4 - 5 | 543 | 110 | ||
| AK080948.1 | - | 4 - 5 | 501 | 110 | ||
| AK140343.1 | - | 4 - 5 | 503 | 110 | ||
| BC079671.1 | - | 4 - 5 | 527 | 30532682 | 110 | |
| L23971.1 | - | 4 - 5 | 388 | 110 | ||
Set Membership: |
|