Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00484502_s1
          See other FRAT1 GE Assays ›
          Gene Symbol:
          Frat1
          Gene Name:
          frequently rearranged in advanced T cell lymphomas
          Gene Aliases:
          AW060382
          Chromosome Location:
          Chr.19: 41829970 - 41832583 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00484502_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Frat1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008043.3 NP_032069.2 1 1893 64
          GenBank mRNA AK164926.1 - 1 589 64
          U58974.2 - 1 1743 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000598.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Mus musculus Mouse Wnt Panel 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Frat1

          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic axis specification
          protein export from nucleus
          Wnt signaling pathway
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32117381 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32117385 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32117383 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32117378 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32117376 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P584188 U58974.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P584188 NM_008043.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P584188 AK164926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P584188 U58974.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P584188 NM_008043.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P584188 AK164926.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179096 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P584188 U58974.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P584188 NM_008043.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P584188 AK164926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P584188 U58974.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P584188 NM_008043.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P584188 AK164926.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P119633 U58974.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P119633 NM_008043.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P119633 AK164926.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32117395 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179096 - -
          Whole Mouse Genome G4122A A_51_P119633 U58974.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119633 NM_008043.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119633 AK164926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P584188 U58974.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P584188 NM_008043.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P584188 AK164926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P584188 U58974.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P584188 NM_008043.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P584188 AK164926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P504524 U58974.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P504524 NM_008043.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P504524 AK015680.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P504524 BC140987.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118915 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179096 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14331977 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14331979 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32117383 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32117381 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32117385 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32117378 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32117376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32117434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14358198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14362234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14331977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14362231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14431222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14224955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14374298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14377232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14344147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14484496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14431214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32117434 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10463167 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463166 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463165 - -
          Affymetrix HT MG-430 PM Array Plate 1449814_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10463164 - -
          Mouse Genome 430 2.0 Array 1449814_PM_at - -
          Mouse Genome 430 2.0 Array 1449814_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14296_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14296_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900005355.mm.1 NM_008043.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005355.mm.1 NM_008043.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005355.mm.1 TC1900000598.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005361.mm.1 NM_008043.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005361.mm.1 NM_008043.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005361.mm.1 TC1900000598.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005356.mm.1 NM_008043.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005356.mm.1 NM_008043.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005356.mm.1 TC1900000598.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005357.mm.1 NM_008043.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005357.mm.1 NM_008043.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005357.mm.1 TC1900000598.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005359.mm.1 NM_008043.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005359.mm.1 NM_008043.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005359.mm.1 TC1900000598.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005358.mm.1 NM_008043.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005358.mm.1 NM_008043.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005358.mm.1 TC1900000598.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005360.mm.1 NM_008043.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005360.mm.1 NM_008043.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005360.mm.1 TC1900000598.mm.1 MTA TC
          Mouse Expression Set 430 1449814_at - -
          Mouse Expression Set 430 1449814_PM_at - -
          Murine Genome U74v2 Set 92695_at - -
          Murine Genome U74v2 Set 92695_PM_at - -
          Mouse Genome 430A 2.0 Array 1449814_PM_at - -
          Mouse Genome 430A 2.0 Array 1449814_at - -
          Affymetrix HT Mouse Genome 430A Array 1449814_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449814_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 866410 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2120731 - -
          Illumina mouse-6 v1.0 expression beadchip 780520 - -
          Illumina mouse-6 v1.0 expression beadchip 2120731 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780520 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120731 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227593 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2737584 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227593 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2737584 - -
          mouse-6 v1.1 (Illumina) scl000436.1_94-S - -
          mouse-6 v1.1 (Illumina) scl53126.1.80_140-S - -
          Illumina Sentrix mouse-6.v1 bead array 780520 - -
          Illumina Sentrix mouse-6.v1 bead array 2120731 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53126.1.80_140-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000436.1_94-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120731 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780520 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1251022 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1227593 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227593 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251022 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline