Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00484518_s1
          See other FUT1 GE Assays ›
          Gene Symbol:
          Fut1
          Gene Name:
          fucosyltransferase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 45617266 - 45621059 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00484518_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fut1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271981.1 NP_001258910.1 2 854 86
          NM_008051.6 NP_032077.2 2 992 86
          GenBank mRNA AF113533.1 - 1 848 86
          AK137638.1 - 2 994 86
          BC109145.1 - 2 830 40045410 86
          BC109146.1 - 2 830 40045419 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000855.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fut1

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          fucosylation
          galactoside 2-alpha-L-fucosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986247 AK137638.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986247 NM_008051.5 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 101888 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136269 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986247 AK137638.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986247 NM_008051.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P222604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25094061 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136269 - -
          Whole Mouse Genome G4122A A_51_P222604 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133820 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106087 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136269 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06323704 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06497469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25093990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25094114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06010420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06010427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06497467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06323719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06323710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06010425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25094070 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10553030 - -
          Affymetrix Mouse Gene 1.0 ST Array 10553028 - -
          Affymetrix Mouse Gene 1.0 ST Array 10553029 - -
          Affymetrix HT MG-430 PM Array Plate 1450359_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10553027 - -
          Mouse Genome 430 2.0 Array 1450359_PM_at - -
          Mouse Genome 430 2.0 Array 1450359_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14343_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14343_at - -
          Affymetrix Mouse Clariom S Array PSR0700007460.mm.1 NM_001271981.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007460.mm.1 NM_008051.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007460.mm.1 TC0700000855.mm.1 MTA TC
          Mouse Expression Set 430 1450359_PM_at - -
          Mouse Expression Set 430 1450359_at - -
          Murine Genome U74v2 Set 99381_at - -
          Murine Genome U74v2 Set 99381_PM_at - -
          Mouse Genome 430A 2.0 Array 1450359_at - -
          Mouse Genome 430A 2.0 Array 1450359_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450359_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450359_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 645075 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1500068 - -
          Illumina mouse-6 v1.0 expression beadchip 1500068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500068 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2744081 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2744081 - -
          mouse-6 v1.1 (Illumina) scl014343.2_310-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl014343.2_310-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2777013 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2777013 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline