Search
Search

B4galnt2| Gene Symbol: B4galnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008081.3 | NP_032107.1 | 3 - 4 | 404 | 105 | |
| GenBank mRNA | BC071231.1 | - | 3 - 4 | 372 | 105 | |
| BC139165.1 | - | 3 - 4 | 394 | 8862187 | 105 | |
| BC139166.1 | - | 3 - 4 | 394 | 8862188 | 105 | |
| L30104.1 | - | 3 - 4 | 404 | 105 | ||
Set Membership: |
|