Search
Search

Sypl2| Gene Symbol: Sypl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008596.1 | NP_032622.1 | 3 - 4 | 437 | 73 | |
| GenBank mRNA | AB010140.1 | - | 3 - 4 | 437 | 73 | |
| AK052749.1 | - | 3 - 4 | 483 | 73 | ||
| AK082471.1 | - | 3 - 4 | 456 | 73 | ||
| AK083422.1 | - | 3 - 4 | 487 | 73 | ||
| AK162663.1 | - | 3 - 4 | 467 | 73 | ||
| BC030038.1 | - | 3 - 4 | 420 | 1068035 | 73 | |
Set Membership: |
|