Search
Search

Meis3| Gene Symbol: Meis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK083907.1 | - | 6 - 7 | 1043 | 133 | |
| AK134758.1 | - | 7 - 8 | 815 | 133 | ||
| AK155364.1 | - | 7 - 8 | 827 | 133 | ||
| BC003762.1 | - | 7 - 8 | 818 | 3593200 | 133 | |
| BC117532.1 | - | 7 - 8 | 790 | 40106322 | 133 | |
| BC118513.1 | - | 7 - 8 | 790 | 40106324 | 133 | |
| BI987529.1 | - | 4 - 5 | 280 | 133 | ||
| U57344.1 | - | 7 - 8 | 845 | 133 | ||
Set Membership: |
|