Search
Search

Rad54l| Gene Symbol: Rad54l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122958.1 | NP_001116430.1 | 13 - 14 | 1668 | 78 | |
| NM_001122959.1 | NP_001116431.1 | 13 - 14 | 1647 | 78 | ||
| NM_009015.3 | NP_033041.3 | 12 - 13 | 1998 | 78 | ||
| GenBank mRNA | AK088876.1 | - | 13 - 14 | 1665 | 78 | |
| AK161644.1 | - | 13 - 14 | 1621 | 78 | ||
| BC021643.1 | - | 12 - 13 | 1979 | 3987790 | 78 | |
| X97796.1 | - | 12 - 13 | 1458 | 78 | ||
Set Membership: |
|