Search
Search

Sptbn1| Gene Symbol: Sptbn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009260.2 | NP_033286.2 | 29 - 30 | 6630 | 77 | |
| NM_175836.2 | NP_787030.2 | 29 - 30 | 6290 | 77 | ||
| GenBank mRNA | AF017112.1 | - | 29 - 30 | 6537 | 77 | |
| AK051630.1 | - | 4 - 5 | 648 | 77 | ||
| AK161119.1 | - | 8 - 9 | 1413 | 77 | ||
| AK220456.1 | - | 30 - 31 | 6492 | 77 | ||
| M74773.1 | - | 29 - 30 | 6610 | 77 | ||
Set Membership: |
|