Search
Search

Capn2| Gene Symbol: Capn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009794.3 | NP_033924.2 | 18 - 19 | 2036 | 75 | |
| GenBank mRNA | AF015038.1 | - | 18 - 19 | 1982 | 75 | |
| AK052141.1 | - | 16 - 17 | 1625 | 75 | ||
| AK150695.1 | - | 18 - 19 | 2010 | 75 | ||
| AK152094.1 | - | 18 - 19 | 2010 | 75 | ||
| AK152790.1 | - | 18 - 19 | 2013 | 75 | ||
| AK152994.1 | - | 18 - 19 | 2013 | 75 | ||
| AK154855.1 | - | 18 - 19 | 2050 | 75 | ||
| BC054726.1 | - | 18 - 19 | 2036 | 5703563 | 75 | |
| D38117.1 | - | 18 - 19 | 2077 | 75 | ||
| Y10139.1 | - | 18 - 19 | 1918 | 75 | ||
Set Membership: |
|