Search
Search

Cbfa2t3| Gene Symbol: Cbfa2t3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109873.1 | NP_001103343.1 | 8 - 9 | 1363 | 62 | |
| NM_009824.2 | NP_033954.2 | 8 - 9 | 1344 | 62 | ||
| NM_177289.1 | NP_796263.1 | 7 - 8 | 1285 | 62 | ||
| GenBank mRNA | AF038029.1 | - | 8 - 9 | 1226 | 62 | |
| AK033680.1 | - | 7 - 8 | 1181 | 62 | ||
| AK041887.1 | - | 8 - 9 | 1366 | 62 | ||
| BC141396.1 | - | 8 - 9 | 1173 | 9056009 | 62 | |
| BC151156.1 | - | 8 - 9 | 1173 | 9088044 | 62 | |
Set Membership: |
|