Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00486938_m1
          See other CDH5 GE Assays ›
          Gene Symbol:
          Cdh5
          Gene Name:
          cadherin 5
          Gene Aliases:
          7B4, AA408225, Cd144, VE-Cad, VECD, VEcad, Vec
          Chromosome Location:
          Chr.8: 104101613 - 104144504 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00486938_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdh5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009868.4 NP_033998.2 7 - 8 1341 69
          GenBank mRNA AK164474.1 - 7 - 8 1324 69
          AK167620.1 - 7 - 8 1273 69
          BC054790.1 - 7 - 8 1267 6831962 69
          D63942.1 - 7 - 8 1302 69
          X83930.2 - 7 - 8 1256 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001176.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Stem Cell Pluripotency Inventoried Mouse_Stem_Cell Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdh5

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel maturation
          cell adhesion
          homophilic cell adhesion via plasma membrane adhesion molecules
          negative regulation of cell proliferation
          single organismal cell-cell adhesion
          regulation of establishment of cell polarity
          receptor binding
          calcium ion binding
          protein binding
          beta-catenin binding
          protein phosphatase binding
          ion channel binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800009136.mm.1 NM_009868.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009136.mm.1 NM_009868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009136.mm.1 TC0800001176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009142.mm.1 NM_009868.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009142.mm.1 NM_009868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009142.mm.1 TC0800001176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009135.mm.1 NM_009868.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009135.mm.1 NM_009868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009135.mm.1 TC0800001176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009137.mm.1 NM_009868.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009137.mm.1 NM_009868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009137.mm.1 TC0800001176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009133.mm.1 NM_009868.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009133.mm.1 NM_009868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009133.mm.1 TC0800001176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009131.mm.1 NM_009868.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009131.mm.1 NM_009868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009131.mm.1 TC0800001176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009138.mm.1 NM_009868.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009138.mm.1 NM_009868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009138.mm.1 TC0800001176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009140.mm.1 NM_009868.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009140.mm.1 NM_009868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009140.mm.1 TC0800001176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009132.mm.1 NM_009868.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009132.mm.1 NM_009868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009132.mm.1 TC0800001176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009141.mm.1 NM_009868.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009141.mm.1 NM_009868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009141.mm.1 TC0800001176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009134.mm.1 NM_009868.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009134.mm.1 NM_009868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009134.mm.1 TC0800001176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009139.mm.1 NM_009868.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009139.mm.1 NM_009868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009139.mm.1 TC0800001176.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009143.mm.1 NM_009868.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009143.mm.1 NM_009868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009143.mm.1 TC0800001176.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0