Search
Search

Mthfr| Gene Symbol: Mthfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161798.1 | NP_001155270.1 | 8 - 9 | 1487 | 132 | |
| NM_010840.3 | NP_034970.2 | 8 - 9 | 1410 | 132 | ||
| NR_027809.1 | 8 - 9 | 1487 | 132 | |||
| GenBank mRNA | AK030192.1 | - | 8 - 9 | 1405 | 132 | |
| AK149795.1 | - | 8 - 9 | 1472 | 132 | ||
| AK150159.1 | - | 8 - 9 | 1488 | 132 | ||
| BC051017.1 | - | 8 - 9 | 1389 | 6308248 | 132 | |
| BC052466.1 | - | 8 - 9 | 1454 | 6834886 | 132 | |
Set Membership: |
|