Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00488845_m1
          See other RACGAP1 GE Assays ›
          Gene Symbol:
          Racgap1
          Gene Name:
          Rac GTPase-activating protein 1
          Gene Aliases:
          AI227039, AI327394, Band25, GTPase, MgcRacGAP, gtl11, mKIAA1478
          Chromosome Location:
          Chr.15: 99620496 - 99651656 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00488845_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Racgap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001253808.1 NP_001240737.1 8 - 9 905 95
          NM_001253809.1 NP_001240738.1 8 - 9 901 95
          NM_012025.7 NP_036155.1 9 - 10 1036 95
          GenBank mRNA AB030252.1 - 8 - 9 885 95
          AF079974.1 - 8 - 9 846 95
          AF212320.1 - 8 - 9 872 95
          AF212321.1 - 9 - 10 1003 95
          AK144608.1 - 9 - 10 1004 95
          AK144745.1 - 8 - 9 865 95
          AK154929.1 - 9 - 10 1024 95
          AK162660.1 - 6 - 7 4105 95
          AK165897.1 - 8 - 9 858 95
          AK166084.1 - 9 - 10 994 95
          AK166094.1 - 8 - 9 863 95
          AK168020.1 - 8 - 9 862 95
          AK168025.1 - 8 - 9 862 95
          AK168170.1 - 8 - 9 862 95
          AK168679.1 - 8 - 9 846 95
          AK214098.1 - 4 - 5 335 95
          BC010715.1 - 8 - 9 851 3602242 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002217.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Racgap1

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          actomyosin contractile ring assembly
          transport
          ion transport
          cell cycle
          signal transduction
          multicellular organism development
          spermatogenesis
          sulfate transport
          embryo development
          cell differentiation
          positive regulation of cytokinesis
          intracellular signal transduction
          positive regulation of GTPase activity
          mitotic spindle midzone assembly
          cell division
          regulation of attachment of spindle microtubules to kinetochore
          mitotic spindle assembly
          GTPase activator activity
          phosphatidylinositol-3,4,5-trisphosphate binding
          microtubule binding
          lipid binding
          protein kinase binding
          alpha-tubulin binding
          gamma-tubulin binding
          metal ion binding
          beta-tubulin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500018166.mm.1 NM_001253808.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018166.mm.1 NM_001253808.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018166.mm.1 NM_001253809.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018166.mm.1 NM_001253809.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018166.mm.1 NM_012025.7.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018166.mm.1 NM_012025.7 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018166.mm.1 TC1500002217.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline