Search
Search

Cpsf2| Gene Symbol: Cpsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016856.3 | NP_058552.1 | 6 - 7 | 855 | 61 | |
| GenBank mRNA | AF012822.1 | - | 6 - 7 | 841 | 61 | |
| AK054372.1 | - | 6 - 7 | 838 | 61 | ||
| AK140869.1 | - | 6 - 7 | 837 | 61 | ||
| AK141325.1 | - | 6 - 7 | 837 | 61 | ||
| AK147746.1 | - | 6 - 7 | 856 | 61 | ||
| BC007163.1 | - | 4 - 5 | 513 | 61 | ||
| BC013628.1 | - | 6 - 7 | 830 | 3497854 | 61 | |
Set Membership: |
|