Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00489780_m1
          See other CNOT4 GE Assays ›
          Gene Symbol:
          Cnot4
          Gene Name:
          CCR4-NOT transcription complex, subunit 4
          Gene Aliases:
          Not4, Not4h, Not4hp
          Chromosome Location:
          Chr.6: 35022065 - 35133737 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00489780_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cnot4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164411.1 NP_001157883.1 1 - 2 237 83
          NM_001164412.1 NP_001157884.1 1 - 2 237 83
          NM_001164413.1 NP_001157885.1 1 - 2 237 83
          NM_001164414.1 NP_001157886.1 1 - 2 237 83
          NM_016877.4 NP_058573.3 1 - 2 237 83
          GenBank mRNA AK009234.1 - 1 - 2 217 83
          AK028190.1 - 1 - 2 107 84
          AK029753.1 - 1 - 2 205 83
          AK032248.1 - 1 - 2 217 83
          AK075887.1 - 1 - 2 225 83
          AK144760.1 - 1 - 2 195 83
          AK165997.1 - 1 - 2 223 83
          BC048243.1 - 1 - 2 125 6493673 83
          BC058778.1 - 1 - 2 189 5397231 83
          U71269.1 - 1 - 2 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002059.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK009234.1,AK028190.1,AK032248.1,AK165997.1,BC048243.1,BC058778.1,NM_001164411.1,NM_001164412.1,NM_001164413.1,NM_001164414.1,NM_016877.4,U71269.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cnot4

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein autoubiquitination
          nucleotide binding
          nucleic acid binding
          RNA binding
          ubiquitin-protein transferase activity
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          poly(A) RNA binding
          metal ion binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600016249.mm.1 NM_001164411.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016249.mm.1 NM_001164411.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016249.mm.1 NM_001164412.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016249.mm.1 NM_001164412.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016249.mm.1 NM_001164413.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016249.mm.1 NM_001164413.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016249.mm.1 NM_001164414.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016249.mm.1 NM_001164414.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016249.mm.1 NM_016877.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016249.mm.1 NM_016877.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016249.mm.1 TC0600002059.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600016250.mm.1 NM_001164411.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016250.mm.1 NM_001164411.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016250.mm.1 NM_001164412.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016250.mm.1 NM_001164412.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016250.mm.1 NM_001164413.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016250.mm.1 NM_001164413.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016250.mm.1 NM_001164414.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016250.mm.1 NM_001164414.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016250.mm.1 NM_016877.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016250.mm.1 NM_016877.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016250.mm.1 TC0600002059.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600016251.mm.1 NM_001164411.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016251.mm.1 NM_001164411.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016251.mm.1 NM_001164412.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016251.mm.1 NM_001164412.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016251.mm.1 NM_001164413.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016251.mm.1 NM_001164413.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016251.mm.1 NM_001164414.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016251.mm.1 NM_001164414.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016251.mm.1 NM_016877.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016251.mm.1 NM_016877.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016251.mm.1 TC0600002059.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600016252.mm.1 NM_001164411.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016252.mm.1 NM_001164411.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016252.mm.1 NM_001164412.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016252.mm.1 NM_001164412.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016252.mm.1 NM_001164413.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016252.mm.1 NM_001164413.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016252.mm.1 NM_001164414.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016252.mm.1 NM_001164414.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016252.mm.1 NM_016877.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016252.mm.1 NM_016877.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600016252.mm.1 TC0600002059.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 866075 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline