Search
Search
| Gene Symbol: Srpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016911.4 | NP_058607.1 | 6 - 7 | 897 | 101 | |
| GenBank mRNA | AB028049.1 | - | 6 - 7 | 824 | 101 | |
| AB028050.1 | - | 4 - 5 | 572 | 101 | ||
| AK034696.1 | - | 6 - 7 | 899 | 101 | ||
| AK159662.1 | - | 5 - 6 | 885 | 101 | ||
| BC022572.1 | - | 6 - 7 | 853 | 3993025 | 101 | |
Set Membership: |
|