Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00490401_g1
          See other CTS8 GE Assays ›
          Gene Symbol:
          Cts8
          Gene Name:
          cathepsin 8
          Gene Aliases:
          CTS2, Epcs68, Epcs70
          Chromosome Location:
          Chr.13: 61246747 - 61255348 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00490401_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cts8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019541.3 NP_062414.3 3 - 4 331 109
          GenBank mRNA AF250840.1 - 3 - 4 343 109
          AK049474.1 - 3 - 4 332 109
          AY014780.1 - 3 - 4 330 109
          BC068241.1 - 3 - 4 321 30019120 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002149.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cts8

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel remodeling
          proteolysis
          protein catabolic process
          proteolysis involved in cellular protein catabolic process
          trophoblast giant cell differentiation
          cysteine-type endopeptidase activity
          peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P11137 BC068241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P11137 NM_019541.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P11137 AK049474.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620680 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 42011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 644766 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61492 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125363 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P11137 BC068241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P11137 NM_019541.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P11137 AK049474.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279473 BC068241.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279473 NM_019541.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279473 AF250840.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279473 AK049474.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29089122 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125363 - -
          Whole Mouse Genome G4122A A_51_P279473 BC068241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279473 NM_019541.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279473 AF250840.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279473 AK049474.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118266 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131224 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125363 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10765583 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10799905 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10765590 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10765594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29089033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10765594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10842981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11117635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10715402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10715405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11092633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10842979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10583017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29089088 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10409928 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409930 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409932 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409931 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409927 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409933 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409929 - -
          Affymetrix HT MG-430 PM Array Plate 1446266_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420680_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10409924 - -
          Mouse Genome 430 2.0 Array 1420680_at - -
          Mouse Genome 430 2.0 Array 1446266_PM_at - -
          Mouse Genome 430 2.0 Array 1446266_at - -
          Mouse Genome 430 2.0 Array 1420680_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56094_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56094_at - -
          Affymetrix Mouse Clariom S Array PSR1300013821.mm.1 NM_019541.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013821.mm.1 NM_019541.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013821.mm.1 TC1300002149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013824.mm.1 NM_019541.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013824.mm.1 NM_019541.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013824.mm.1 TC1300002149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013822.mm.1 NM_019541.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013822.mm.1 NM_019541.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013822.mm.1 TC1300002149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013823.mm.1 NM_019541.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013823.mm.1 NM_019541.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013823.mm.1 TC1300002149.mm.1 MTA TC
          Mouse Expression Set 430 1420680_at - -
          Mouse Expression Set 430 1420680_PM_at - -
          Mouse Expression Set 430 1446266_PM_at - -
          Mouse Expression Set 430 1446266_at - -
          Mouse Genome 430A 2.0 Array 1420680_PM_at - -
          Mouse Genome 430A 2.0 Array 1420680_at - -
          Affymetrix HT Mouse Genome 430A Array 1420680_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420680_at - -
          Affymetrix HT Mouse Genome 430B Array 1446266_at - -
          Affymetrix HT Mouse Genome 430B Array 1446266_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 392585 - -
          ABI Mouse Genome Survey Microarray 824064 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5340139 - -
          Illumina mouse-6 v1.0 expression beadchip 5340139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005340139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2640538 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2640538 - -
          mouse-6 v1.1 (Illumina) scl43857.8_139-S - -
          Illumina Sentrix mouse-6.v1 bead array 5340139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43857.8_139-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5340139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2824793 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2640538 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2824793 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2640538 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline