Search
Search

Cdh17| Gene Symbol: Cdh17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019753.4 | NP_062727.1 | 7 - 8 | 926 | 81 | |
| GenBank mRNA | AF177669.1 | - | 7 - 8 | 822 | 81 | |
| AK089791.1 | - | 7 - 8 | 893 | 81 | ||
| AK155633.1 | - | 7 - 8 | 927 | 81 | ||
| AK171733.1 | - | 7 - 8 | 849 | 81 | ||
| BC042891.1 | - | 7 - 8 | 844 | 4208360 | 81 | |
| D87912.1 | - | 7 - 8 | 871 | 81 | ||
Set Membership: |
|