Search
Search

Mmp19| Gene Symbol: Mmp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021412.2 | NP_067387.1 | 1 - 2 | 192 | 116 | |
| GenBank mRNA | AF153199.1 | - | 1 - 2 | 144 | 116 | |
| AF155221.1 | - | 1 - 2 | 172 | 116 | ||
| AF162446.1 | - | 1 - 2 | 172 | 116 | ||
| AK154863.1 | - | 1 - 2 | 185 | 116 | ||
| BC112966.2 | - | 1 - 2 | 164 | 40039023 | 116 | |
Set Membership: |
|