| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600024138.mm.1 |
NM_007881.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024138.mm.1 |
NM_007881.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024138.mm.1 |
TC0600003604.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600024135.mm.1 |
NM_007881.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024135.mm.1 |
NM_007881.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024135.mm.1 |
TC0600003604.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600024136.mm.1 |
NM_007881.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024136.mm.1 |
NM_007881.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600024136.mm.1 |
TC0600003604.mm.1 |
MTA TC |