Search
Search
| Gene Symbol: Ggt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079235.1 | - | 11 - 12 | 1584 | 63 | |
| BC012969.1 | - | 10 - 11 | 1472 | 4037243 | 63 | |
| BU756502.1 | - | 3 - 4 | 63 | |||
| CT010196.1 | - | 9 - 10 | 1330 | 63 | ||
| U30509.1 | - | 10 - 11 | 1490 | 63 | ||
Set Membership: |
|