Search
Search

Icam5| Gene Symbol: Icam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008319.2 | NP_032345.2 | 5 - 6 | 1310 | 99 | |
| GenBank mRNA | AK135040.1 | - | 5 - 6 | 1223 | 99 | |
| BC113140.1 | - | 5 - 6 | 1258 | 40048010 | 99 | |
| U06483.1 | - | 5 - 6 | 1258 | 99 | ||
Set Membership: |
|