Search
Search
| Gene Symbol: Lbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008489.2 | NP_032515.2 | 13 - 14 | 1505 | 87 | |
| GenBank mRNA | BC004795.1 | - | 13 - 14 | 1393 | 3589072 | 87 |
| BQ176753.1 | - | 2 - 3 | 87 | |||
| X99347.1 | - | 13 - 14 | 1455 | 87 | ||
Set Membership: |
|