Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00493434_m1
          See other HNF1A GE Assays ›
          Gene Symbol:
          Hnf1a
          Gene Name:
          HNF1 homeobox A
          Gene Aliases:
          AI323641, HNF1, HNF1-alpha, HNF1[a], Hnf-1, Hnf1alpha, LFB1, Tcf-1, Tcf1
          Chromosome Location:
          Chr.5: 114948980 - 114971062 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00493434_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hnf1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009327.3 NP_033353.2 1 - 2 543 134
          GenBank mRNA AK141225.1 - 1 - 2 350 134
          AK144051.1 - 1 - 2 550 134
          BC080698.1 - 1 - 2 545 30471380 134
          M57966.1 - 523 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003084.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hnf1a

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          blastocyst development
          liver development
          placenta development
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          fatty acid biosynthetic process
          bile acid biosynthetic process
          heme biosynthetic process
          response to oxidative stress
          protein localization
          cholesterol metabolic process
          response to glucose
          bile acid and bile salt transport
          fatty acid transport
          insulin secretion
          regulation of Wnt signaling pathway
          embryonic limb morphogenesis
          endocrine pancreas development
          renal glucose absorption
          glucose homeostasis
          reverse cholesterol transport
          bone resorption
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          glucose import
          regulation of hormone secretion
          paraxial mesoderm formation
          reproductive structure development
          regulation of insulin secretion
          positive regulation of transcription initiation from RNA polymerase II promoter
          SMAD protein signal transduction
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription cofactor binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          protein homodimerization activity
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity
          protein dimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500027424.mm.1 TC0500003084.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027424.mm.1 NM_009327.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027424.mm.1 NM_009327.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027426.mm.1 TC0500003084.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027426.mm.1 NM_009327.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027426.mm.1 NM_009327.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027427.mm.1 TC0500003084.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027427.mm.1 NM_009327.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027427.mm.1 NM_009327.3 Refseq

          Back To Top
           

          Citations (1)

          Fibroblast growth factor 21 corrects obesity in mice.
          Authors
          Coskun T, Bina HA, Schneider MA, Dunbar JD, Hu CC, Chen Y, Moller DE, Kharitonenkov A
          Journal
          Endocrinology - Page No 6018 (Mon Dec 01 00:00:00 EST 2008)
          Volume (Year)
          149 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline