Search
Search

Unc5c| Gene Symbol: Unc5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293561.1 | NP_001280490.1 | 14 - 15 | 2494 | 94 | |
| NM_009472.4 | NP_033498.1 | 13 - 14 | 2437 | 94 | ||
| GenBank mRNA | AK031655.1 | - | 14 - 15 | 2461 | 94 | |
| AK138987.1 | - | 2 - 3 | 234 | 94 | ||
| AK141181.1 | - | 14 - 15 | 2460 | 94 | ||
| AK142411.1 | - | 14 - 15 | 2460 | 94 | ||
| AK142974.1 | - | 14 - 15 | 2388 | 94 | ||
| AK164239.1 | - | 13 - 14 | 2238 | 94 | ||
| BC115772.1 | - | 14 - 15 | 2484 | 40085998 | 94 | |
| U72634.1 | - | 13 - 14 | 2437 | 94 | ||
Set Membership: |
|