Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm00494340_g1
See other ZFPM1 GE Assays ›
Gene Symbol:
Zfpm1
Gene Name:
zinc finger protein, multitype 1
Gene Aliases:
FOG, Fog1
Chromosome Location:
Chr.8: 122282141 - 122337862 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm00494340_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Zfpm1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_009569.3
NP_033595.1
7 - 8
1325
63
GenBank mRNA
AF006492.1
-
7 - 8
1329
63
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as MTA transcript cluster(s) TC0800001481.mm.1
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Detect Genomic DNA
Mus musculus
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Zfpm1
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
outflow tract morphogenesis
atrioventricular valve morphogenesis
mitral valve formation
tricuspid valve formation
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
heart development
regulation of definitive erythrocyte differentiation
erythrocyte differentiation
megakaryocyte differentiation
platelet formation
granulocyte differentiation
negative regulation of protein binding
regulation of chemokine production
embryonic hemopoiesis
megakaryocyte development
positive regulation of interferon-gamma biosynthetic process
negative regulation of interleukin-4 biosynthetic process
negative regulation of fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
homeostasis of number of cells
cardiac muscle tissue morphogenesis
definitive erythrocyte differentiation
primitive erythrocyte differentiation
negative regulation of mast cell differentiation
ventricular septum morphogenesis
atrial septum morphogenesis
transcriptional activation by promoter-enhancer looping
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
nucleic acid binding
DNA binding
transcription corepressor activity
protein binding
transcription factor binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR0800012050.mm.1
NM_009569.4
Refseq
Affymetrix Mouse Clariom S Array
PSR0800012050.mm.1
TC0800001481.mm.1
MTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline