| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600026001.mm.1 |
NM_001080552.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026001.mm.1 |
NM_001080552.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600026001.mm.1 |
TC0600003316.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600025998.mm.1 |
NM_001080552.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025998.mm.1 |
NM_001080552.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025998.mm.1 |
NM_001080553.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025998.mm.1 |
NM_001080553.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025998.mm.1 |
NM_010352.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025998.mm.1 |
NM_010352.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025998.mm.1 |
TC0600003316.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600026002.mm.1 |
TC0600003316.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600025999.mm.1 |
NM_001080552.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025999.mm.1 |
NM_001080552.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025999.mm.1 |
NM_001080553.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025999.mm.1 |
NM_001080553.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025999.mm.1 |
NM_010352.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025999.mm.1 |
NM_010352.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025999.mm.1 |
TC0600003316.mm.1 |
MTA TC |