Search
Search
| Gene Symbol: Vnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011704.3 | NP_035834.2 | 3 - 4 | 727 | 92 | |
| GenBank mRNA | AJ132098.1 | - | 3 - 4 | 560 | 92 | |
| AK002773.1 | - | 3 - 4 | 559 | 92 | ||
| AK145984.1 | - | 3 - 4 | 633 | 92 | ||
| AK149485.1 | - | 1 - 2 | 108 | 92 | ||
| AK167427.1 | - | 3 - 4 | 559 | 92 | ||
| BC019203.1 | - | 3 - 4 | 553 | 5041670 | 92 | |
Set Membership: |
|