Search
Search

Ikzf2| Gene Symbol: Ikzf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011770.4 | NP_035900.2 | 6 - 7 | 989 | 105 | |
| GenBank mRNA | AF044257.1 | - | 6 - 7 | 884 | 105 | |
| AK047925.1 | - | 6 - 7 | 851 | 105 | ||
| BC138606.1 | - | 6 - 7 | 829 | 8861626 | 105 | |
| BC138608.1 | - | 6 - 7 | 829 | 8861628 | 105 | |
Set Membership: |
|