Search
Search

Tceb3| Gene Symbol: Tceb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013736.4 | NP_038764.2 | 4 - 5 | 1446 | 116 | |
| GenBank mRNA | AB025015.1 | - | 4 - 5 | 1430 | 116 | |
| AK036592.1 | - | 4 - 5 | 1447 | 116 | ||
| AK145994.1 | - | 4 - 5 | 1447 | 116 | ||
| AK147088.1 | - | 4 - 5 | 1441 | 116 | ||
| AK193358.1 | - | 1 - 2 | 158 | 116 | ||
| BC049885.1 | - | 4 - 5 | 5387774 | 116 | ||
Set Membership: |
|