Search
Search

Abcc2| Gene Symbol: Abcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013806.2 | NP_038834.2 | 30 - 31 | 4406 | 60 | |
| GenBank mRNA | AB008832.1 | - | 2 - 3 | 306 | 60 | |
| AF227274.1 | - | 30 - 31 | 4310 | 60 | ||
| AF282773.1 | - | 30 - 31 | 4310 | 60 | ||
Set Membership: |
|