Search
Search

Incenp| Gene Symbol: Incenp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016692.3 | NP_057901.2 | 1 - 2 | 199 | 56 | |
| GenBank mRNA | AF117610.1 | - | 1 - 2 | 135 | 56 | |
| AK034764.1 | - | 1 - 2 | 128 | 56 | ||
| AK081841.1 | - | 1 - 2 | 133 | 56 | ||
| AK088627.1 | - | 1 - 2 | 156 | 56 | ||
| BC021761.2 | - | 1 - 2 | 96 | 56 | ||
| BC037011.1 | - | 1 - 2 | 152 | 56 | ||
| BC052414.1 | - | 1 - 2 | 132 | 6401188 | 56 | |
| BY788161.1 | - | 1 - 2 | 199 | 56 | ||
Set Membership: |
|