Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00497359_g1
          See other TMOD4 GE Assays ›
          Gene Symbol:
          Tmod4
          Gene Name:
          tropomodulin 4
          Gene Aliases:
          MTMOD, Sk-Tmod
          Chromosome Location:
          Chr.3: 95124117 - 95129208 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00497359_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmod4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016712.3 NP_057921.1 9 - 10 1121 116
          GenBank mRNA AF177174.1 - 8 - 9 1040 116
          AK144408.1 - 9 - 10 1122 116
          AK144535.1 - 9 - 10 1122 116
          BC068020.1 - 8 - 9 1057 30291625 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000889.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmod4

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          actin filament organization
          biological_process
          myofibril assembly
          pointed-end actin filament capping
          actin binding
          tropomyosin binding
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976574 AF177174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976574 AK144408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976574 NM_016712.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976574 AK144535.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976574 BC068020.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767023 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890974 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148619 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111461 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976574 AF177174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976574 AK144408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976574 NM_016712.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976574 AK144535.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976574 BC068020.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P258829 AF177174.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P258829 AK144408.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P258829 NM_016712.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P258829 AK144535.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P258829 BC068020.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22342336 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111461 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148619 - -
          Whole Mouse Genome G4122A A_51_P258829 AF177174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258829 AK144408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258829 NM_016712.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258829 AK144535.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258829 BC068020.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120857 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111461 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148619 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02355578 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02355573 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02355585 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02355571 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02355580 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02355567 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02355576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02355578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02736813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02355575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02355574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02355580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03019069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02355568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22342365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02736809 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10494162 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494163 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494166 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494164 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494161 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494168 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494167 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494169 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494165 - -
          Affymetrix HT MG-430 PM Array Plate 1449969_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10494160 - -
          Mouse Genome 430 2.0 Array 1449969_PM_at - -
          Mouse Genome 430 2.0 Array 1449969_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50874_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50874_at - -
          Affymetrix Mouse Clariom S Array PSR0300006915.mm.1 NM_016712.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006915.mm.1 NM_016712.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006915.mm.1 TC0300000889.mm.1 MTA TC
          Mouse Expression Set 430 1449969_PM_at - -
          Mouse Expression Set 430 1449969_at - -
          Mouse Genome 430A 2.0 Array 1449969_PM_at - -
          Mouse Genome 430A 2.0 Array 1449969_at - -
          Affymetrix HT Mouse Genome 430A Array 1449969_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449969_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 331075 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5910520 - -
          Illumina mouse-6 v1.0 expression beadchip 2360021 - -
          Illumina mouse-6 v1.0 expression beadchip 4590722 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002360021 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005910520 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004590722 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2488846 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2519313 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2438793 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2438793 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2488846 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2519313 - -
          mouse-6 v1.1 (Illumina) scl22890.10.1_51-S - -
          mouse-6 v1.1 (Illumina) scl0001984.1_38-S - -
          mouse-6 v1.1 (Illumina) scl0002067.1_49-S - -
          Illumina Sentrix mouse-6.v1 bead array 2360021 - -
          Illumina Sentrix mouse-6.v1 bead array 4590722 - -
          Illumina Sentrix mouse-6.v1 bead array 5910520 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001984.1_38-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002067.1_49-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22890.10.1_51-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5910520 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2360021 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4590722 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2488846 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2438793 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2519313 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2488846 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2438793 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2519313 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline